ContactUsLinkComponent
Thermometer similar to IP 101 C, solid stem, white backed, 20+150:1°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA341450-FL
Supplier:  Amarell
Thermometer similar to IP 101 C, solid stem, white backed, 20+150:1°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA341450-FL
Supplier:  Amarell
Frequently Bought Together