Thermometer similar to IP 101 C, solid stem, white backed, 20+150:1C, capillary tube specially coated inside, with non-wetting blu e special liquid, immersion 57mm, 290x6-8mm, null
Catalog # AMARA341450-CB
Supplier: Amarell
Frequently Bought Together