ContactUsLinkComponent
Thermometer, solid stem, similar to IP 92 C, white backed, 38,6+41,4:0,05°C, capillary tube specially coated inside, with non-wetti ng blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA341270-CB
Supplier:  Amarell
Thermometer, solid stem, similar to IP 92 C, white backed, 38,6+41,4:0,05°C, capillary tube specially coated inside, with non-wetti ng blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA341270-CB
Supplier:  Amarell
Frequently Bought Together