Flash point-Therm., similar Pensky-Martens, similar DIN, +40+160:1°C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prisma tic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government verif 1 * 1 items
Catalog # AMARM230044-FL
Supplier: Amarell
Frequently Bought Together