Hydrometer DIN 12791/BS 718, series L 50 SP precision, 1,050-1,100:0,0005g/ml, without therm., ref. temp. 15C, max. 335mm long, sui table for government verification 1 * 1 items
Hydrometer DIN 12791/BS 718, series L 50 SP precision, 1,050-1,100:0,0005g/ml, without therm., ref. temp. 15C, max. 335mm long, sui table for government verification 1 * 1 items