ContactUsLinkComponent
Viscometer-Therm., similar Vogel-Ossag, similar DIN 12785, +70+105:0,1°C, enclosed scale, capillary tube specially coated inside, w ith non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240106-CB
Supplier:  Amarell
Viscometer-Therm., similar Vogel-Ossag, similar DIN 12785, +70+105:0,1°C, enclosed scale, capillary tube specially coated inside, w ith non-wetting blue special liquid, 470mm long, lower part solid stem 200x5mm, 208mm immersion, Richter-top finish, null
Catalog # AMARM240106-CB
Supplier:  Amarell
Frequently Bought Together