Thermometer, solid stem, similar to ASTM 72 F, white backed, -2,5+2,5:0,1F, capillary tube specially coated inside, with non-wetti ng blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA301030-CB
Supplier: Amarell
Frequently Bought Together