Precision therm., high temp., similiar DIN, -10+610:1°C, solid stem, frosted back, total immersion, gallium filled, 450mm long, diam eter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
AMARL29602-GA
: Amarell
Frequently Bought Together