Precision therm., high temp., similar DIN, +200+480:1°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diam eter 6-7mm, durable pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARL29004-GA
Supplier: Amarell
Frequently Bought Together