Precision therm., high temp., similar DIN, -10+480:1°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diame ter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
AMARL29000-GA
: Amarell
Frequently Bought Together