Thermometer similar to AFNOR, STL/0,2/35/85, +35+85:0,2°C, solid stem, white backed, capillary tube specially coated inside, with n on-wetting blue special liquid, 375mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA411038-CB
Supplier: Amarell
Frequently Bought Together