ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 11d, solid stem, white backed, 105+115:0,1°C, length 300mm, imm. 100mm, capillary tube speciall y coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400070-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 11d, solid stem, white backed, 105+115:0,1°C, length 300mm, imm. 100mm, capillary tube speciall y coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400070-CB
Supplier:  Amarell
Frequently Bought Together