Order Entry
Slovenia
Orders LinkContactUsLinkComponent
3-Chlorophenol

3-Chlorophenol

Bottle, 500 g
Catalog # APOSOR8225-500G
Irritant
Environment
Supplier:   Apollo Scientific
3-Chlorophenol
Catalog # APOSOR8225-500G
Supplier:  Apollo Scientific
CAS Number:  108-43-0
3-Chlorophenol

Bottle, 500 g

CAS Number:  108-43-0
Price
203,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:108-43-0
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002256
  • Molecular Weight:128.56 g/mol
  • Boiling Pt:214 °C
  • Melting Pt:30…34 °C
  • Density:1.245 g/cm³ (25 °C)
  • Flash Pt:120 °C
  • Storage Temperature:Ambient
  • EINECS:203-582-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.