3-Chlorophenol
Bottle, 25 g
Catalog # APOSOR8225-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:108-43-0
- Formula:C₆H₅ClO
- MDL Number:MFCD00002256
- Molecular Weight:128.56 g/mol
- Boiling Pt:214 °C
- Melting Pt:30…34 °C
- Density:1.245 g/cm³ (25 °C)
- Flash Pt:120 °C
- Storage Temperature:Ambient
- EINECS:203-582-6
- UN:2020
- ADR:6.1,III
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



