Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

2-Chlorophenol 99%

Bottle, 25 g
Catalog # APOSOR52255-25G
Irritant
Environment
Supplier:   Apollo Scientific
2-Chlorophenol 99%
Catalog # APOSOR52255-25G
Supplier:  Apollo Scientific
CAS Number:  95-57-8
2-Chlorophenol 99%

Bottle, 25 g

CAS Number:  95-57-8
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents

Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).

Synonyms:  1-Chloro-2-hydroxybenzene
  • CAS Number:95-57-8
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002159
  • Molecular Weight:128.56 g/mol
  • Boiling Pt:175…176 °C
  • Melting Pt:7…8 °C
  • Melting Pt:7-8 °C
  • Density:1.26 g/cm³ (25 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-433-2
  • UN:2021
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications