Orders LinkContactUsLinkComponent
2-Chlorophenol 99%
Bottle, 100 g
  APOSOR52255-100G
2-Chlorophenol 99%
  APOSOR52255-100G
 :  95-57-8
2-Chlorophenol 99%
Bottle, 100 g
 :  95-57-8
47,80 €
1 * 100 g
Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).
 :  1-Chloro-2-hydroxybenzene
  •  :95-57-8
  •  :C₆H₅ClO
  •  :MFCD00002159
  •  :128.56 g/mol
  •  :175…176 °C
  •  :7…8 °C
  •  :7-8 °C
  •  :1.26 g/cm³ (25 °C)
  •  :64 °C
  •  :Ambient
  •  :202-433-2
  •  :2021
  •  :6.1,III
  • Pack type:
    Bottle
  • Pk:
    100 g