2-Chlorophenol 99%
Bottle, 100 g
APOSOR52255-100G
Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).
: 1-Chloro-2-hydroxybenzene
- :95-57-8
- :C₆H₅ClO
- :MFCD00002159
- :128.56 g/mol
- :175…176 °C
- :7…8 °C
- :7-8 °C
- :1.26 g/cm³ (25 °C)
- :64 °C
- :Ambient
- :202-433-2
- :2021
- :6.1,III
- Pack type:Bottle
- Pk:100 g