Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Terbium 99,9%, chips

Terbium 99,9%, chips

Bottle, 5 g
Catalog # APOSIN3454-5G
Flammable
Supplier:   Apollo Scientific
Terbium 99,9%, chips
Catalog # APOSIN3454-5G
Supplier:  Apollo Scientific
CAS Number:  7440-27-9
Terbium 99,9%, chips

Bottle, 5 g

CAS Number:  7440-27-9
Price
254,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:7440-27-9
  • Formula:Tb
  • MDL Number:MFCD00011256
  • Molecular Weight:158.93 g/mol
  • Boiling Pt:2800 °C
  • Melting Pt:1356 °C
  • Density:8.234 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3178
  • ADR:4.1,II
  •  :Density: 8.27 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications