Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Boron phosphate 99,9%

Bottle, 100 g
Catalog # APOSIN1296-100G
Irritant
Supplier:   Apollo Scientific
Boron phosphate 99,9%
Catalog # APOSIN1296-100G
Supplier:  Apollo Scientific
CAS Number:  13308-51-5
Boron phosphate 99,9%

Bottle, 100 g

CAS Number:  13308-51-5
Price
222,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:13308-51-5
  • Formula:BO₄P
  • MDL Number:MFCD00011318
  • Molecular Weight:105.78 g/mol
  • Melting Pt:>1400 °C
  • Melting Pt:>1400 °C
  • Density:2.81 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications