Bismuth(III) carbonate basic 99.99%
Bottle, 25 g
APOSIN1239-25G
: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
- :5892-10-4
- :(BiO)₂CO₃
- :MFCD00010877
- :509.97 g/mol
- :6.86 g/cm³ (25 °C)
- :Ambient
- :227-567-9
- Pack type:Bottle
- Pk:25 g