Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Recombinant staphylokinase

1 mg
Catalog # APOSBITP1348-1MG
Supplier:   Apollo Scientific
Recombinant staphylokinase
Catalog # APOSBITP1348-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant staphylokinase

1 mg

CAS Number:  
Price
921,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1348-1MG
  • Enzyme type:
    Recombinant
  • Storage conditions:
    Ambient
  • Enzyme synonyms:
    Staphylokinase
  • MDL No.:
    MFCD09702076
  • Enzyme name:
    Staphylokinase

Specifications