Orders LinkContactUsLinkComponent
α-Cyclodextrin
α-Cyclodextrin
Bottle, 50 g
Catalog # APOSBIM1014-50G
Supplier:   Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIM1014-50G
Supplier:  Apollo Scientific
CAS Number:  10016-20-3
α-Cyclodextrin
Bottle, 50 g
CAS Number:  10016-20-3
Price
123,00 €
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Storage Temperature:Ambient
Specifications
Specifications
  • Pack type:
    Bottle
  • Pk:
    50 g
Specifications