Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99.5%
L(+)-Ascorbic acid ≥99.5%
Bottle, 1 kg
Catalog # APOSBIA0602-1KG
Corrosive
Irritant
Supplier:   Apollo Scientific
L(+)-Ascorbic acid ≥99.5%
Catalog # APOSBIA0602-1KG
Supplier:  Apollo Scientific
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥99.5%
Bottle, 1 kg
CAS Number:  50-81-7
Price
46,40 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Melting Pt:190…194 °C (dec.)
  • Density:1.65 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
Specifications
Specifications
  • Pack type:
    Bottle
  • Pk:
    1 kg