Mop systems, Edgeless®
Standard edgeless mop head, sterile, -, -, -
Catalog # CONTEL-MOPIR
Supplier: CONTEC
Jump to:
Product Details & Documents
Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.
- Mop heads are free from any cellulose contaminants
- Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
- All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation
Mop heads are laundered and packaged in a ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
: All mop system components must be ordered separately. Plastic bucket systems include cart, buckets, wringer, and casters. Stainless steel bucket systems include trolley, buckets, wringer, and casters.
Documents
Specifications
- Capacity:-
- Description:Standard edgeless mop head, sterile
- Dimensions:-
- Length:-
- Seq:01