ContactUsLinkComponent
A leukopak is an enriched leukapheresis product collected from normal peripheral blood (PB). A leukopak contains 85% mononuclear ce lls. PB is collected from healthy volunteer donors under IRB approved informed consent by leukapheresis and tested ne 1 * 1 PAC
  PB001C-3
A leukopak is an enriched leukapheresis product collected from normal peripheral blood (PB). A leukopak contains 85% mononuclear ce lls. PB is collected from healthy volunteer donors under IRB approved informed consent by leukapheresis and tested ne 1 * 1 PAC
  PB001C-3
Frequently Bought Together