CF dye with TCO (trans-cyclooctene) reacts with tetrazine labeled molecule via copper free click chemistry. This copper free bioorth ogonal reaction allows staining the surface of live cells or having concerns about native protein function loss with 1 * 1 mg
Catalog # 96041.
Supplier: Biotium
Frequently Bought Together