ContactUsLinkComponent
Europe norm stackable crate, closed bottom and walls - base 30x20 cm - 5 Liters. Raw material : high density polyethylene. For food use. Colour grey. Inside size 260x160x100 mm 1 * 1 items
Catalog # LAMA648
Supplier:  LAMAPLAST
Europe norm stackable crate, closed bottom and walls - base 30x20 cm - 5 Liters. Raw material : high density polyethylene. For food use. Colour grey. Inside size 260x160x100 mm 1 * 1 items
Catalog # LAMA648
Supplier:  LAMAPLAST
Frequently Bought Together