ContactUsLinkComponent
Safety Container, Material: High density Polyethylene Colour: Yellow. Against leakage and overflow. Suitable for dangerous or polluting liquids. Doesn't require any maintenance. Capacity 300 L. 800 x 1350 x 420 mm. 1 * 1 items
Catalog # LAMA3294
Supplier:  LAMAPLAST
Safety Container, Material: High density Polyethylene Colour: Yellow. Against leakage and overflow. Suitable for dangerous or polluting liquids. Doesn't require any maintenance. Capacity 300 L. 800 x 1350 x 420 mm. 1 * 1 items
Catalog # LAMA3294
Supplier:  LAMAPLAST
Frequently Bought Together