SteamLine combination electrode with screw plug head (ATEX II 1/2G), hot steam sterilizable, CIP and SIP able, glass shaft, SILAMID ® ref., ceramic junction, press. RheoLid ref. system, sensor platinum disk, screw plug head, length 360 mm 1 * 1 items
Catalog # SCOI285113470
Supplier: SI Analytics
Frequently Bought Together