pH combination electrode with screw plug head (ATEX II 1/2G), Low maintenance with Pt 1000, glass shaft, 3 x ceramic junction, gel e lectrolyte, Silamid®-reference system, sphere membrane, A-glass, VP screw plug head, length 225 mm 1 * 1 items
Catalog # SCOI285114820
Supplier: SI Analytics
Frequently Bought Together