DUOLINK INSITU DETECTION REAGENTS ORANGE 1 * 100 Tests
Catalog # SIALDUO92007-100RX
Supplier: Merck
Frequently Bought Together
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
p-Xylene 99% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform ≥99% stabilised ethanol-free
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
DUOLINK INSITU DETECTION REAGENTS ORANGE 1 * 30 Tests
Supplier: Merck
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND