The inner wall of the tube is coated with spray-dried lithium, sodium, or ammonium heparin. these additives are anti-coagulants, whi ch activate anti-thrombins, thus blocking the coagulation cascade of the blood specimen. lh lithium heparin tubes are 1 * 1.200 items
Catalog # GREI454046_1200
Supplier: Greiner Bio-One
Frequently Bought Together