Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 10×/0,25 (WD=17,4 mm), 17,4 mm
Catalog # MOTI1101001705211
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705211
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 10×/0,25 (WD=17,4 mm), 17,4 mm
Price
67,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    10x/0,25
  • Description:
    CCIS® Plan achromatic objective UC PL 10×/0,25 (WD=17,4 mm)
  • Seq:
    03
  • Working distance:
    17,4 mm