Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic Phase objective UC PL Ph 10×/0,25 (WD=17,4 mm), Ph1, 17,4 mm
Catalog # MOTI1101001706481
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001706481
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic Phase objective UC PL Ph 10×/0,25 (WD=17,4 mm), Ph1, 17,4 mm
Price
206,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    Ph1 10x/0,25
  • Description:
    CCIS® Plan achromatic Phase objective UC PL Ph 10×/0,25 (WD=17,4 mm), Ph1
  • Seq:
    09
  • Working distance:
    17,4 mm