Orders LinkContactUsLinkComponent
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 4×/0,1 (WD=30,5 mm), 30,5 mm
Catalog # MOTI1101001705201
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705201
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 4×/0,1 (WD=30,5 mm), 30,5 mm
Price
128,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    4x/0,1
  • Description:
    CCIS® Plan achromatic objective UC PL 4×/0,1 (WD=30,5 mm)
  • Seq:
    02
  • Working distance:
    30,5 mm