ContactUsLinkComponent
Capillary, coated, Zero Flow brand, covalently bonded, neutral charge, 100um ID by 1meter. Use in Capillary Electrophoresis. 1/pack. 1 * 1 m
Catalog # MICS04600-ZF
Supplier:  MICROSOLV
undefined
Capillary, coated, Zero Flow brand, covalently bonded, neutral charge, 100um ID by 1meter. Use in Capillary Electrophoresis. 1/pack. 1 * 1 m
Catalog # MICS04600-ZF
Supplier:  MICROSOLV
Frequently Bought Together