Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
50* ml, 14/23, 140 ± 5 mm, 0,080 ± ml
  HIRS2820378
 :   HIRSCHMANN
Volumetric flasks, class A, blue markings
  HIRS2820378
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
50* ml, 14/23, 140 ± 5 mm, 0,080 ± ml
45,90 €
1 * 2 items
DURAN®, borosilicate glass, without stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
 : DIN EN ISO 1042
  • NS:
    14/23
  • Capacity:
    50* ml
  • Height:
    140 ± 5 mm
  • Seq:
    18
  • Tolerance:
    0,080 ± ml