Volumetric flasks, class A, blue markings
100 ml, 12/21, 170 ± 5 mm, 0,100 ± ml
Catalog # HIRS2820380
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, without stopper, conformity certified (DE-M marking).
- Blue enamelled indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certifications: DIN EN ISO 1042
Documents
Specifications
- NS:12/21
- Capacity:100 ml
- Height:170 ± 5 mm
- Seq:20
- Tolerance:0,100 ± ml



