ContactUsLinkComponent
MULTIPARAMETER TRANSM MIQ/TC2020XT-3G C6 Multi parameter Controller to connect up to 20 IQ sensors. Scope of delivery: MIQ/TC 2020 3G Terminal/Controller, MIQ/C6 module with 6 current outputs (0/4…20 mA), MIQ/PS wide range power supply 1 * 1 items
Catalog # WTWO470024
Supplier:  WTW ONLINE
MULTIPARAMETER TRANSM MIQ/TC2020XT-3G C6 Multi parameter Controller to connect up to 20 IQ sensors. Scope of delivery: MIQ/TC 2020 3G Terminal/Controller, MIQ/C6 module with 6 current outputs (0/4…20 mA), MIQ/PS wide range power supply 1 * 1 items
Catalog # WTWO470024
Supplier:  WTW ONLINE
Frequently Bought Together