Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
250 ml, 14/23, 0,15 ± ml
Catalog # POUL1.5125504F
Supplier:   Poulten & Graf
Volumetric flasks, class A, blue markings
Catalog # POUL1.5125504F
Supplier:  Poulten & Graf
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
250 ml, 14/23, 0,15 ± ml
Price
29,90 €
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate glass 3.3 with PP stopper.
  • Calibrated to contain (TC, In)
  • With ground ring mark
  • Conformity approved
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    250 ml
  • Seq:
    20
  • Tolerance:
    0,15 ± ml