Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Catalog # POUL1.5125304F
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass 3.3 with PP stopper.
- Calibrated to contain (TC, In)
- With ground ring mark
- Conformity approved
Certifications: DIN EN ISO 1042
Specifications
- NS:14/23
- Capacity:200 ml
- Seq:19
- Tolerance:0,15 ± ml