Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Catalog # POUL1.5125304F
Supplier:   Poulten & Graf
Volumetric flasks, class A, blue markings
Catalog # POUL1.5125304F
Supplier:  Poulten & Graf
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Price
28,70 €
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate glass 3.3 with PP stopper.
  • Calibrated to contain (TC, In)
  • With ground ring mark
  • Conformity approved
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    200 ml
  • Seq:
    19
  • Tolerance:
    0,15 ± ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.