ContactUsLinkComponent
HT100 instrument with full touchscreen colour graphic display. Includes PK19M Penetration probe, Protective Boot and Micro USB cable. Free software via weblink download, see Quick Reference Guide supplied when ordered. -73°C to +537°C / -99°F to +999 1 * 1 items
Catalog # COMAHT100/PK19
Supplier:  COMARK
HT100 instrument with full touchscreen colour graphic display. Includes PK19M Penetration probe, Protective Boot and Micro USB cable. Free software via weblink download, see Quick Reference Guide supplied when ordered. -73°C to +537°C / -99°F to +999 1 * 1 items
Catalog # COMAHT100/PK19
Supplier:  COMARK
Frequently Bought Together