ContactUsLinkComponent
with ‘’Connect Assist’’, ceramic shaft and integral solvent trap ring for use with cup CCB25 DIN for TM-PE-C and TM-LI-C32. Material: Titanium 1 * 1 items
Catalog # HAAK222-2125
Supplier:  HAAKE
with ‘’Connect Assist’’, ceramic shaft and integral solvent trap ring for use with cup CCB25 DIN for TM-PE-C and TM-LI-C32. Material: Titanium 1 * 1 items
Catalog # HAAK222-2125
Supplier:  HAAKE
Frequently Bought Together