ContactUsLinkComponent
elma tec clean A5 Suitable for: metal, artificial and natural rubber Suitable for: metal, artificial and natural rubber surfaces also suitable for gray cast iron and zinc die casting as well as for light metals. Removes scaled or resinous oil and gr 1 * 850 g
Catalog # ELMA8000141
Supplier:  ELMA SCHMIDBAUER
elma tec clean A5 Suitable for: metal, artificial and natural rubber Suitable for: metal, artificial and natural rubber surfaces also suitable for gray cast iron and zinc die casting as well as for light metals. Removes scaled or resinous oil and gr 1 * 850 g
Catalog # ELMA8000141
Supplier:  ELMA SCHMIDBAUER
Frequently Bought Together