ContactUsLinkComponent
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession XP005570 011.1). 1 * 100 µG
Catalog # ACRBLA3-C5252100UG
Supplier:  ACRO Biosystems
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession XP005570 011.1). 1 * 100 µG
Catalog # ACRBLA3-C5252100UG
Supplier:  ACRO Biosystems
Frequently Bought Together