ContactUsLinkComponent
BENCHTOP FREEZE DRYER, including: Condenser in stainless steel to -55C (for room temperature up to 21C) Vacuum pump 8,1 m3/h, with exhaust filter and system of oil recovery PLC smart control with touch screen with capacity to issue lyophilisation r 1 * 1 items
Catalog # TELR61653-1517468
BENCHTOP FREEZE DRYER, including: Condenser in stainless steel to -55C (for room temperature up to 21C) Vacuum pump 8,1 m3/h, with exhaust filter and system of oil recovery PLC smart control with touch screen with capacity to issue lyophilisation r 1 * 1 items
Catalog # TELR61653-1517468
Frequently Bought Together