ContactUsLinkComponent
PolyGard®-2 Sensor Cartridge, 5VDC Kohlendioxid CO2:0-5% VOL (Infrarotsensor) zum Anschluss an SB2/MSB2/MSC2, digitale Kalibration, Schutzart:IP65 1 * 1 ST
Catalog # SNSTSC2-I1164-B
PolyGard®-2 Sensor Cartridge, 5VDC Kohlendioxid CO2:0-5% VOL (Infrarotsensor) zum Anschluss an SB2/MSB2/MSC2, digitale Kalibration, Schutzart:IP65 1 * 1 ST
Catalog # SNSTSC2-I1164-B
Frequently Bought Together