PRESSURE JACKET FOR HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSEL MADE OF CERAMIC FOR TITAN MPS™ 8 POSITION MICROWAVE SAMPLE PREPA RATION SYSTEMS. 1 * 100 items
PRESSURE JACKET FOR HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSEL MADE OF CERAMIC FOR TITAN MPS™ 8 POSITION MICROWAVE SAMPLE PREPA RATION SYSTEMS. 1 * 100 items