ContactUsLinkComponent
UNCALIBRATED 99% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281920
Supplier:  PerkinElmer
UNCALIBRATED 99% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 58 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 60 MM SPINNER. 1 * 1 items
Catalog # PERCL1281920
Supplier:  PerkinElmer
Frequently Bought Together