ContactUsLinkComponent
THE GLADIATR VISION™ IS A SAMPLING TOOL WHICH COUPLES SMALL AREA INFRARED ANALYSIS WITH SIMULTANEOUS VIEWING. SAMPLES ARE PLACED FAC E DOWN AND POSITIONED ON THE DIAMOND CRYSTAL WHILE ITS IMAGE IS PROJECTED IN REAL-TIME ON THE LCD SCREEN. FINDING AND 1 * 1 items
Catalog # PERCL1272423
Supplier:  PerkinElmer
THE GLADIATR VISION™ IS A SAMPLING TOOL WHICH COUPLES SMALL AREA INFRARED ANALYSIS WITH SIMULTANEOUS VIEWING. SAMPLES ARE PLACED FAC E DOWN AND POSITIONED ON THE DIAMOND CRYSTAL WHILE ITS IMAGE IS PROJECTED IN REAL-TIME ON THE LCD SCREEN. FINDING AND 1 * 1 items
Catalog # PERCL1272423
Supplier:  PerkinElmer
Frequently Bought Together