VACUUM MODEL MV-1CR(BP) S/S BACKPACK 240V FROM TIGER-VAC -INCLUDES BASIC ACCESSORIES Technical Features: Volt: 240V, Hertz: 50Hz, Watt: 1000, Ampere: 4.2A. Dry recovery. Air Flow: 187 m3/h. Vacuum Pressure: 1753 mmH2O Recovery capacity: Dry: with Pa 1 * 1 items
TIGV110737D
Frequently Bought Together